siRNA Details
| Gene Symbol: Wdr59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170742.1 | NP_001164213.1 | 18, 19 | 1985 | ||
| NM_176923.4 | NP_795897.2 | 18, 19 | 1985 | |||
| GenBank mRNA | AK030644.1 | - | 18, 19 | 1866 | ||
| AK040807.1 | - | 3, 4 | 272 | |||
| AK136303.1 | - | 14, 15 | 1537 | |||
| AK170910.1 | - | 18, 19 | 1984 | |||
| BC066082.1 | - | 18, 19 | 1970 | 30355918 | ||
| EF011622.1 | - | 18, 19 | 1970 | |||

Wdr59
Genomic Map