siRNA Details
| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009261.3 | NP_033287.2 | 17 | 8375 | ||
| GenBank mRNA | AK032246.1 | - | 1 | 1999 | ||
| AK045193.1 | - | 1 | 1284 | |||
| AY380798.1 | - | 1 | 2001 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Strbp| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009261.3 | NP_033287.2 | 17 | 8375 | ||
| GenBank mRNA | AK032246.1 | - | 1 | 1999 | ||
| AK045193.1 | - | 1 | 1284 | |||
| AY380798.1 | - | 1 | 2001 | |||