siRNA Details
| Gene Symbol: Idi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145360.2 | NP_663335.2 | 3, 4 | 662 | ||
| GenBank mRNA | AK029302.1 | - | 3, 4 | 295 | ||
| AK084208.1 | - | 3, 4 | 292 | |||
| AK132073.1 | - | 3, 4 | 296 | |||
| AK160832.1 | - | 3, 4 | 296 | |||
| BC004801.1 | - | 2, 3 | 283 | 3589498 | ||
| BC110313.1 | - | 3, 4 | 300 | 5343431 | ||
| CT010310.1 | - | 2, 3 | 240 | |||

Idi1
Genomic Map