siRNA Details
| Gene Symbol: Idi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145360.2 | NP_663335.2 | 2, 3 | 562 | ||
| GenBank mRNA | AK029302.1 | - | 2, 3 | 195 | ||
| AK084208.1 | - | 2, 3 | 192 | |||
| AK132073.1 | - | 2, 3 | 195 | |||
| AK160832.1 | - | 2, 3 | 196 | |||
| BC004801.1 | - | 1, 2 | 183 | 3589498 | ||
| BC110313.1 | - | 2, 3 | 200 | 5343431 | ||
| CT010310.1 | - | 1, 2 | 140 | |||

Idi1
Genomic Map