siRNA Details
| Gene Symbol: Zfp944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176962.4 | NP_795936.2 | 6 | 847 | ||
| GenBank mRNA | AK031839.1 | - | 6 | 770 | ||
| BC026813.1 | - | 5 | 604 | |||
| BC108345.1 | - | 7 | 863 | 4935739 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp944| Gene Symbol: Zfp944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176962.4 | NP_795936.2 | 6 | 847 | ||
| GenBank mRNA | AK031839.1 | - | 6 | 770 | ||
| BC026813.1 | - | 5 | 604 | |||
| BC108345.1 | - | 7 | 863 | 4935739 | ||