siRNA Details
| Gene Symbol: Galm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176963.4 | NP_795937.1 | 2 | 513 | ||
| GenBank mRNA | AK040998.1 | - | 2 | 514 | ||
| AK085153.1 | - | 2 | 314 | |||
| AK158120.1 | - | 2 | 312 | |||
| AK161687.1 | - | 2 | 312 | |||
| AK170014.1 | - | 2 | 312 | |||
| BC028818.1 | - | 2 | 316 | 5135570 | ||

Galm
Genomic Map