siRNA Details
| Gene Symbol: Galm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176963.4 | NP_795937.1 | 4 | 813 | ||
| GenBank mRNA | AK040998.1 | - | 4 | 814 | ||
| AK158120.1 | - | 4 | 612 | |||
| AK161687.1 | - | 4 | 612 | |||
| AK170014.1 | - | 4 | 614 | |||
| BC028818.1 | - | 4 | 616 | 5135570 | ||

Galm
Genomic Map