siRNA Details
| Gene Symbol: Fsd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195284.1 | NP_001182213.1 | 3 | 293 | ||
| NM_176966.4 | NP_795940.1 | 3 | 293 | |||
| GenBank mRNA | AK038893.1 | - | 3 | 271 | ||
| AK052914.1 | - | 3 | 336 | |||
| AK140020.1 | - | 3 | 277 | |||
| BB662298.1 | - | 3 | 331 | |||
| BC130265.1 | - | 3 | 209 | 40142990 | ||

Fsd1l
Genomic Map