siRNA Details
| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010568.2 | NP_034698.2 | 21 | 6290 | ||
| GenBank mRNA | AK081856.1 | - | 1 | 616 | ||
| AK085344.1 | - | 1 | 866 | |||
| AK166467.1 | - | 1 | 1112 | |||
| AK205131.1 | - | 1 | 38 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Insr| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010568.2 | NP_034698.2 | 21 | 6290 | ||
| GenBank mRNA | AK081856.1 | - | 1 | 616 | ||
| AK085344.1 | - | 1 | 866 | |||
| AK166467.1 | - | 1 | 1112 | |||
| AK205131.1 | - | 1 | 38 | |||