siRNA Details
| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010568.2 | NP_034698.2 | 21 | 6168 | ||
| GenBank mRNA | AK081856.1 | - | 1 | 494 | ||
| AK085344.1 | - | 1 | 744 | |||
| AK166467.1 | - | 1 | 990 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Insr| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010568.2 | NP_034698.2 | 21 | 6168 | ||
| GenBank mRNA | AK081856.1 | - | 1 | 494 | ||
| AK085344.1 | - | 1 | 744 | |||
| AK166467.1 | - | 1 | 990 | |||