siRNA Details
| Gene Symbol: Eml5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081191.1 | NP_001074660.1 | 38 | 5500 | ||
| GenBank mRNA | AK044024.1 | - | 2 | 153 | ||
| BC027154.1 | - | 1 | 45 | |||
| BC158137.1 | - | 36 | 4972 | 9007401 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eml5| Gene Symbol: Eml5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081191.1 | NP_001074660.1 | 38 | 5500 | ||
| GenBank mRNA | AK044024.1 | - | 2 | 153 | ||
| BC027154.1 | - | 1 | 45 | |||
| BC158137.1 | - | 36 | 4972 | 9007401 | ||