siRNA Details
| Gene Symbol: Ankar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176980.4 | NP_795954.2 | 7 | 1730 | ||
| GenBank mRNA | AK030065.1 | - | 7 | 1731 | ||
| BC132415.1 | - | 7 | 1705 | 40130692 | ||
| BC145253.1 | - | 7 | 1705 | 9053805 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankar| Gene Symbol: Ankar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176980.4 | NP_795954.2 | 7 | 1730 | ||
| GenBank mRNA | AK030065.1 | - | 7 | 1731 | ||
| BC132415.1 | - | 7 | 1705 | 40130692 | ||
| BC145253.1 | - | 7 | 1705 | 9053805 | ||