siRNA Details
| Gene Symbol: Ankar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176980.4 | NP_795954.2 | 3 | 905 | ||
| GenBank mRNA | AK030065.1 | - | 3 | 906 | ||
| AK077036.1 | - | 2 | 285 | |||
| BC132415.1 | - | 3 | 880 | 40130692 | ||
| BC145253.1 | - | 3 | 880 | 9053805 | ||

Ankar
Genomic Map