siRNA Details
Gene Symbol: Frmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028127.3 | NP_082403.2 | 4 | 506 | ||
GenBank mRNA | BC053929.1 | - | 4 | 506 | 6389695 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Frmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028127.3 | NP_082403.2 | 4 | 506 | ||
GenBank mRNA | BC053929.1 | - | 4 | 506 | 6389695 |