siRNA Details
| Gene Symbol: Simc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176987.4 | NP_795961.3 | 2 | 2549 | ||
| GenBank mRNA | AK052561.1 | - | 1 | 343 | ||
| AK166176.1 | - | 1 | 153 | |||
| BC059035.1 | - | 1 | 808 | |||
| BC094550.1 | - | 1 | 360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Simc1| Gene Symbol: Simc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176987.4 | NP_795961.3 | 2 | 2549 | ||
| GenBank mRNA | AK052561.1 | - | 1 | 343 | ||
| AK166176.1 | - | 1 | 153 | |||
| BC059035.1 | - | 1 | 808 | |||
| BC094550.1 | - | 1 | 360 | |||