siRNA Details
| Gene Symbol: Cacna2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033382.2 | NP_001028554.2 | 29 | 2794 | ||
| GenBank mRNA | AK137847.1 | - | 3 | 100 | ||
| BC141091.1 | - | 28 | 2650 | 9055703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacna2d4| Gene Symbol: Cacna2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033382.2 | NP_001028554.2 | 29 | 2794 | ||
| GenBank mRNA | AK137847.1 | - | 3 | 100 | ||
| BC141091.1 | - | 28 | 2650 | 9055703 | ||