siRNA Details
| Gene Symbol: Igf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183029.2 | NP_898850.2 | 1, 2 | 520 | ||
| GenBank mRNA | AK049196.1 | - | 1, 2 | 217 | ||
| AK077404.1 | - | 1, 2 | 199 | |||
| AK159654.1 | - | 1, 2 | 255 | |||
| AY531659.1 | - | 1, 2 | 184 | |||
| BC023758.1 | - | 1, 2 | 218 | |||
| BC054552.1 | - | 1, 2 | 240 | |||
| CB290059.1 | - | 1, 2 | 520 | |||

Igf2bp2
Genomic Map