siRNA Details
| Gene Symbol: Slc22a30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177002.3 | NP_795976.1 | 7 | 1378 | ||
| GenBank mRNA | AK040781.1 | - | 7 | 1381 | ||
| AK050438.1 | - | 7 | 1380 | |||
| AK052592.1 | - | 8 | 1476 | |||
| AK080180.1 | - | 7 | 1380 | |||
| BC132146.1 | - | 7 | 1316 | 40130423 | ||
| BC132336.1 | - | 7 | 1316 | 40130613 | ||

Slc22a30
Genomic Map