siRNA Details
| Gene Symbol: Tmem229a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177013.3 | NP_795987.2 | 1 | 2199 | ||
| GenBank mRNA | AK032121.1 | - | 1 | 209 | ||
| AK087465.1 | - | 1 | 2167 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem229a| Gene Symbol: Tmem229a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177013.3 | NP_795987.2 | 1 | 2199 | ||
| GenBank mRNA | AK032121.1 | - | 1 | 209 | ||
| AK087465.1 | - | 1 | 2167 | |||