siRNA Details
| Gene Symbol: Bbs9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178415.1 | NP_848502.1 | 19 | 2384 | ||
| NM_181316.4 | NP_851833.2 | 18 | 2328 | |||
| GenBank mRNA | AK028200.1 | - | 18 | 2418 | ||
| AK053503.1 | - | 3 | 1073 | |||
| AK086114.1 | - | 18 | 2300 | |||
| AK144730.1 | - | 19 | 2355 | |||
| AK166297.1 | - | 18 | 2309 | |||
| BC030419.1 | - | 16 | 1692 | |||
| BC046426.1 | - | 18 | 2340 | 3601505 | ||
| BC054441.1 | - | 7 | 587 | |||

Bbs9
Genomic Map