siRNA Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027225.1 | NP_081501.1 | 2 | 427 | ||
| NM_177025.5 | NP_795999.2 | 2 | 427 | |||
| GenBank mRNA | AK037010.1 | - | 3 | 418 | ||
| AK052533.1 | - | 2 | 376 | |||
| AK052939.1 | - | 3 | 358 | |||
| AK083510.1 | - | 3 | 379 | |||
| AK144943.1 | - | 3 | 471 | |||
| AK164624.1 | - | 3 | 357 | |||
| AK173076.1 | - | 2 | 258 | |||
| AY308746.1 | - | 2 | 427 | |||
| BC067007.1 | - | 2 | 424 | 5702930 | ||

Cobll1
Genomic Map