siRNA Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027225.1 | NP_081501.1 | 11 | 1962 | ||
| NM_177025.5 | NP_795999.2 | 12 | 2073 | |||
| GenBank mRNA | AK037010.1 | - | 13 | 2067 | ||
| AK052533.1 | - | 11 | 1911 | |||
| AK135859.1 | - | 3 | 301 | |||
| AK144943.1 | - | 13 | 2120 | |||
| AK173076.1 | - | 11 | 1793 | |||
| AY308746.1 | - | 12 | 2073 | |||
| BC067007.1 | - | 11 | 1959 | 5702930 | ||

Cobll1
Genomic Map