siRNA Details
| Gene Symbol: Dock6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177030.3 | NP_796004.2 | 28 | 3353 | ||
| GenBank mRNA | AK129348.1 | - | 25 | 3321 | ||
| BC057368.1 | - | 8 | 979 | |||
| BU701914.1 | - | 6 | 756 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dock6| Gene Symbol: Dock6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177030.3 | NP_796004.2 | 28 | 3353 | ||
| GenBank mRNA | AK129348.1 | - | 25 | 3321 | ||
| BC057368.1 | - | 8 | 979 | |||
| BU701914.1 | - | 6 | 756 | |||