siRNA Details
| Gene Symbol: Dsel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081316.1 | NP_001074785.1 | 2 | 4761 | ||
| GenBank mRNA | AK032110.1 | - | 1 | 2653 | ||
| AK164854.1 | - | 3 | 2061 | |||
| BC120567.1 | - | 1 | 3123 | 40129490 | ||
| BC137796.1 | - | 1 | 3123 | 8860815 | ||

Dsel
Genomic Map