siRNA Details
Gene Symbol: Ism1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276489.1 | NP_001263418.1 | 2, 3 | 862 | ||
GenBank mRNA | AK077398.1 | - | 2, 3 | 863 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ism1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276489.1 | NP_001263418.1 | 2, 3 | 862 | ||
GenBank mRNA | AK077398.1 | - | 2, 3 | 863 |