siRNA Details
| Gene Symbol: Flad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177041.3 | NP_796015.2 | 3 | 892 | ||
| GenBank mRNA | AK044501.1 | - | 901 | |||
| AK161929.1 | - | 3 | 936 | |||
| BC025817.1 | - | 3 | 845 | 5099127 | ||
| BC046769.1 | - | 3 | 892 | 6809196 | ||

Flad1
Genomic Map