siRNA Details
| Gene Symbol: Auts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177047.3 | NP_796021.2 | 2 | 928 | ||
| GenBank mRNA | AK052418.1 | - | 2 | 990 | ||
| AK077988.1 | - | 2 | 1385 | |||
| AK147478.1 | - | 1927 | ||||
| BC096021.1 | - | 2 | 927 | |||
| BC151031.1 | - | 2 | 942 | |||
| BC151047.1 | - | 2 | 941 | |||

Auts2
Genomic Map