siRNA Details
Gene Symbol: Kif6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177052.3 | NP_796026.2 | 9, 10 | 1146 | ||
GenBank mRNA | AK043404.1 | - | 9, 10 | 1103 | ||
AK083760.1 | - | 9, 10 | 1183 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kif6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177052.3 | NP_796026.2 | 9, 10 | 1146 | ||
GenBank mRNA | AK043404.1 | - | 9, 10 | 1103 | ||
AK083760.1 | - | 9, 10 | 1183 |