siRNA Details
| Gene Symbol: Casc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205369.1 | NP_001192298.1 | 5 | 867 | ||
| NM_001205370.1 | NP_001192299.1 | 5 | 867 | |||
| NM_001205371.1 | NP_001192300.1 | 5 | 457 | |||
| NM_177054.5 | NP_796028.2 | 5 | 867 | |||
| NM_199038.3 | NP_950239.2 | 5 | 867 | |||
| GenBank mRNA | AK131751.1 | - | 5 | 458 | ||
| AK160708.1 | - | 5 | 708 | |||
| AY484583.1 | - | 5 | 707 | |||
| AY484585.1 | - | 5 | 707 | |||
| BC034556.1 | - | 5 | 730 | |||
| BC064446.1 | - | 5 | 867 | 6843743 | ||

Casc4
Genomic Map