siRNA Details
| Gene Symbol: Tmem198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177056.4 | NP_796030.1 | 5 | 1411 | ||
| GenBank mRNA | AK038957.1 | - | 5 | 1372 | ||
| AK082334.1 | - | 4 | 1671 | |||
| AK158863.1 | - | 5 | 1885 | |||
| BC042776.1 | - | 3 | 308 | |||
| BC046546.1 | - | 3 | 308 | |||
| BC059868.1 | - | 5 | 1376 | 6809137 | ||

Tmem198
Genomic Map