siRNA Details
| Gene Symbol: Sidt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159419.1 | NP_001152891.1 | 19 | 2265 | ||
| NM_198034.3 | NP_932151.2 | 19 | 2265 | |||
| GenBank mRNA | AK034344.1 | - | 4 | 415 | ||
| AK046836.1 | - | 19 | 2625 | |||
| AK171920.1 | - | 19 | 2418 | |||
| AY562213.1 | - | 8 | 809 | |||
| AY562214.1 | - | 8 | 809 | |||
| BC025888.1 | - | 9 | 915 | 5039343 | ||
| BC079584.1 | - | 19 | 2265 | 5704672 | ||

Sidt1
Genomic Map