siRNA Details
| Gene Symbol: Sidt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159419.1 | NP_001152891.1 | 8 | 1180 | ||
| NM_198034.3 | NP_932151.2 | 8 | 1180 | |||
| GenBank mRNA | AK046836.1 | - | 8 | 1540 | ||
| AK171920.1 | - | 8 | 1333 | |||
| BC079584.1 | - | 8 | 1180 | 5704672 | ||

Sidt1
Genomic Map