siRNA Details
| Gene Symbol: Ccdc79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_180958.3 | NP_851289.2 | 17 | 2039 | ||
| GenBank mRNA | AB775896.1 | - | 15 | 1817 | ||
| AK029754.1 | - | 17 | 2040 | |||
| AK030157.1 | - | 16 | 2353 | |||
| BC113771.1 | - | 14 | 2079 | |||
| BC129814.1 | - | 15 | 2247 | |||

Ccdc79
Genomic Map