siRNA Details
| Gene Symbol: Ano4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277188.1 | NP_001264117.1 | 21 | 2447 | ||
| GenBank mRNA | BC065114.1 | - | 20 | 2404 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ano4| Gene Symbol: Ano4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277188.1 | NP_001264117.1 | 21 | 2447 | ||
| GenBank mRNA | BC065114.1 | - | 20 | 2404 | ||