siRNA Details
| Gene Symbol: Rps6kc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178775.4 | NP_848890.3 | 10, 11 | 1343 | ||
| GenBank mRNA | AK044809.1 | - | 7, 8 | 1062 | ||
| AK049003.1 | - | 3, 4 | 291 | |||
| AK149858.1 | - | 10, 11 | 1308 | |||
| BC058403.1 | - | 1, 2 | 42 | 6812243 | ||

Rps6kc1
Genomic Map