siRNA Details
| Gene Symbol: Adrbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285806.1 | NP_001272735.1 | 4 | 394 | ||
| NM_177078.4 | NP_796052.2 | 4 | 355 | |||
| GenBank mRNA | AK048763.1 | - | 4 | 320 | ||
| AK076533.1 | - | 4 | 253 | |||
| AK134672.1 | - | 4 | 332 | |||
| AK141186.1 | - | 4 | 372 | |||
| AK172513.1 | - | 4 | 307 | |||
| BY289705.1 | - | 4 | 337 | |||
| CJ104587.1 | - | 4 | 356 | |||

Adrbk2
Genomic Map