siRNA Details
| Gene Symbol: Adrbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285806.1 | NP_001272735.1 | 2 | 234 | ||
| NM_177078.4 | NP_796052.2 | 2 | 234 | |||
| GenBank mRNA | AK048763.1 | - | 2 | 199 | ||
| AK076533.1 | - | 2 | 132 | |||
| AK134672.1 | - | 2 | 211 | |||
| AK141186.1 | - | 2 | 212 | |||
| AK172513.1 | - | 2 | 186 | |||
| CJ104587.1 | - | 2 | 235 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012897, XM_008769317.

Adrbk2
Genomic Map