siRNA Details
| Gene Symbol: BC049715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178776.3 | NP_848891.2 | 3 | 495 | ||
| GenBank mRNA | AK044622.1 | - | 2 | 1710 | ||
| BC049715.1 | - | 3 | 495 | 6741857 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC049715| Gene Symbol: BC049715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178776.3 | NP_848891.2 | 3 | 495 | ||
| GenBank mRNA | AK044622.1 | - | 2 | 1710 | ||
| BC049715.1 | - | 3 | 495 | 6741857 | ||