siRNA Details
| Gene Symbol: Zmat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277239.1 | NP_001264168.1 | 2, 3 | 246 | ||
| NM_177086.4 | NP_796060.1 | 2, 3 | 246 | |||
| GenBank mRNA | AK036254.1 | - | 2, 3 | 245 | ||
| AK134793.1 | - | 2, 3 | 245 | |||
| AK140391.1 | - | 247 | ||||
| AK140413.1 | - | 2, 3 | 247 | |||
| BC065110.1 | - | 2, 3 | 234 | 30536600 | ||
| BC079657.1 | - | 2, 3 | 233 | 6853622 | ||

Zmat4
Genomic Map