siRNA Details
| Gene Symbol: Cep95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166685.1 | NP_001160157.1 | 13, 14 | 1659 | ||
| NM_177088.3 | NP_796062.2 | 14, 15 | 1788 | |||
| GenBank mRNA | AK076400.1 | - | 14, 15 | 1766 | ||
| BC090675.1 | - | 14, 15 | 1744 | |||
| BC116308.1 | - | 13, 14 | 1625 | 40095242 | ||
| BC116309.1 | - | 13, 14 | 1642 | |||

Cep95
Genomic Map