siRNA Details
| Gene Symbol: Fndc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165965.1 | NP_001159437.1 | 5 | 924 | ||
| NM_177091.5 | NP_796065.2 | 6 | 1182 | |||
| GenBank mRNA | AK053989.1 | - | 6 | 1045 | ||
| AK054241.1 | - | 6 | 1182 | |||
| AK165075.1 | - | 3 | 737 | |||
| BC111898.1 | - | 5 | 731 | 40061374 | ||
| BC113188.1 | - | 6 | 988 | |||

Fndc7
Genomic Map