siRNA Details
| Gene Symbol: Lrrc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177093.3 | NP_796067.2 | 3 | 830 | ||
| GenBank mRNA | AK049274.1 | - | 3 | 831 | ||
| AK147818.1 | - | 3 | 806 | |||
| BC141007.1 | - | 3 | 790 | 9055619 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc58| Gene Symbol: Lrrc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177093.3 | NP_796067.2 | 3 | 830 | ||
| GenBank mRNA | AK049274.1 | - | 3 | 831 | ||
| AK147818.1 | - | 3 | 806 | |||
| BC141007.1 | - | 3 | 790 | 9055619 | ||