siRNA Details
| Gene Symbol: Hook3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207659.3 | NP_997542.1 | 19, 20 | 2054 | ||
| GenBank mRNA | AK042180.1 | - | 6, 7 | 538 | ||
| AK084594.1 | - | 2, 3 | 151 | |||
| AK137723.1 | - | 9, 10 | 790 | |||
| AK145149.1 | - | 19, 20 | 2024 | |||
| AK170858.1 | - | 19, 20 | 1999 | |||
| AY223806.1 | - | 19, 20 | 1844 | |||
| BC090960.1 | - | 20, 21 | 1970 | 30651062 | ||

Hook3
Genomic Map