siRNA Details
| Gene Symbol: Hook3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207659.3 | NP_997542.1 | 14 | 1585 | ||
| GenBank mRNA | AK042180.1 | - | 1 | 69 | ||
| AK054263.1 | - | 14 | 1504 | |||
| AK137723.1 | - | 4 | 321 | |||
| AK145149.1 | - | 14 | 1555 | |||
| AK170858.1 | - | 14 | 1530 | |||
| AY223806.1 | - | 14 | 1375 | |||
| BC090960.1 | - | 14 | 1495 | 30651062 | ||

Hook3
Genomic Map