siRNA Details
| Gene Symbol: Maats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081025.1 | NP_001074494.1 | 1 | 191 | ||
| GenBank mRNA | AF503943.1 | - | 1 | 474 | ||
| AK045121.1 | - | 1 | 143 | |||
| AK077038.1 | - | 1 | 177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Maats1| Gene Symbol: Maats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081025.1 | NP_001074494.1 | 1 | 191 | ||
| GenBank mRNA | AF503943.1 | - | 1 | 474 | ||
| AK045121.1 | - | 1 | 143 | |||
| AK077038.1 | - | 1 | 177 | |||