siRNA Details
| Gene Symbol: Ccdc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177111.3 | NP_796085.3 | 9 | 1162 | ||
| GenBank mRNA | AK145297.1 | - | 8 | 1175 | ||
| AK158666.1 | - | 7 | 680 | |||
| BC070471.1 | - | 9 | 1162 | 30626499 | ||
| BC072583.1 | - | 157 | 6403859 | |||

Ccdc66
Genomic Map