siRNA Details
| Gene Symbol: March3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177115.2 | NP_796089.1 | 3 | 662 | ||
| GenBank mRNA | AK036622.1 | - | 3 | 630 | ||
| AK041088.1 | - | 3 | 662 | |||
| BC107232.1 | - | 3 | 446 | 40052073 | ||
| BC107233.1 | - | 3 | 446 | 40052074 | ||

March3
Genomic Map