siRNA Details
| Gene Symbol: 1520401A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028064.1 | - | 24 | 930 | ||
| AK047830.1 | - | 25 | 2054 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1520401A03Rik| Gene Symbol: 1520401A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028064.1 | - | 24 | 930 | ||
| AK047830.1 | - | 25 | 2054 | |||