siRNA Details
| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045615.1 | - | 1 | 501 | ||
| AK054352.1 | - | 1 | 511 | |||
| BC147259.1 | - | 1 | 100 | |||
| BC147260.1 | - | 1 | 100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgea5| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045615.1 | - | 1 | 501 | ||
| AK054352.1 | - | 1 | 511 | |||
| BC147259.1 | - | 1 | 100 | |||
| BC147260.1 | - | 1 | 100 | |||