siRNA Details
| Gene Symbol: PTPRO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002848.3 | NP_002839.1 | 22 | 3556 | ||
| NM_030667.2 | NP_109592.1 | 23 | 3640 | |||
| NM_030668.2 | NP_109593.1 | 10 | 1147 | |||
| NM_030669.2 | NP_109594.1 | 11 | 1231 | |||
| NM_030670.2 | NP_109595.1 | 10 | 1147 | |||
| NM_030671.2 | NP_109596.1 | 11 | 1231 | |||
| GenBank mRNA | AF152378.1 | - | 11 | 1223 | ||
| AF187042.1 | - | 10 | 1139 | |||
| AF187043.1 | - | 11 | 1223 | |||
| AF187044.1 | - | 10 | 1139 | |||
| AK289784.1 | - | 23 | 3385 | |||
| AK290982.1 | - | 10 | 870 | |||
| AK299529.1 | - | 8 | 606 | |||
| BC126201.1 | - | 23 | 3340 | 8991917 | ||
| BC126203.1 | - | 23 | 3340 | 8991919 | ||
| U20489.1 | - | 22 | 3258 | |||
| Z48541.1 | - | 23 | 3290 | |||

PTPRO
Genomic Map